missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-N-Acetyl-α-allylglycine, 98%, 98% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441185000

 View more versions of this product

Catalog No. AC441185000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(R)-N-Acetyl-α-allylglycine
Authentic
White to Yellow
500mg
121786-40-1
100.0
C7H11NO3
QTNLDKHXFVSKCF-ZCFIWIBFSA-N
(2R)-2-acetamidopent-4-enoic acid
41097732
Powder
98.0% min.
-8.3
101°C to 106°C
98% ee
97.5
98%
2r-2-acetamidopent-4-enoic acid, ambotzaaa1953, r, ?-2-acetylamino-4-pentenoic acid, r-n-acetyl-alpha-allylglycine ee, r-n-alpha-acetyl-allylglycineee, rac-2r-2-acetamidopent-4-enoic acid
CC(=O)NC(CC=C)C(=O)O
157.17
157.17
98%,98% ee

Chemical Identifiers

121786-40-1
157.17
2r-2-acetamidopent-4-enoic acid, ambotzaaa1953, r, ?-2-acetylamino-4-pentenoic acid, r-n-acetyl-alpha-allylglycine ee, r-n-alpha-acetyl-allylglycineee, rac-2r-2-acetamidopent-4-enoic acid
(2R)-2-acetamidopent-4-enoic acid
C7H11NO3
QTNLDKHXFVSKCF-ZCFIWIBFSA-N
41097732
CC(=O)NC(CC=C)C(=O)O
SDS Documents