missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-N-Acetyl-α-allylglycine, 98%, 98% ee, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441181000
Specifications
(R)-N-Acetyl-α-allylglycine | |
Authentic | |
White to Yellow | |
100mg | |
121786-40-1 | |
100.0 | |
C7H11NO3 | |
QTNLDKHXFVSKCF-ZCFIWIBFSA-N | |
(2R)-2-acetamidopent-4-enoic acid | |
41097732 | |
Powder |
98.0% min. | |
-8.3 | |
101°C to 106°C | |
98% ee | |
97.5 | |
98% | |
2r-2-acetamidopent-4-enoic acid, ambotzaaa1953, r, ?-2-acetylamino-4-pentenoic acid, r-n-acetyl-alpha-allylglycine ee, r-n-alpha-acetyl-allylglycineee, rac-2r-2-acetamidopent-4-enoic acid | |
CC(=O)NC(CC=C)C(=O)O | |
157.17 | |
157.17 | |
98%,98% ee |
Chemical Identifiers
121786-40-1 | |
157.17 | |
2r-2-acetamidopent-4-enoic acid, ambotzaaa1953, r, ?-2-acetylamino-4-pentenoic acid, r-n-acetyl-alpha-allylglycine ee, r-n-alpha-acetyl-allylglycineee, rac-2r-2-acetamidopent-4-enoic acid | |
(2R)-2-acetamidopent-4-enoic acid |
C7H11NO3 | |
QTNLDKHXFVSKCF-ZCFIWIBFSA-N | |
41097732 | |
CC(=O)NC(CC=C)C(=O)O |