missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-N-Acetyl-α-allylglycine, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

CAS 121786-40-1
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C7H11NO3
Formula Weight 157.17
Synonym (2R)-2-(Acetylamino)-4-pentenoic acid
Chemical Name or Material (R)-N-Acetyl-α-allylglycine, 98% ee
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441181000 SDS Thermo Fisher Scientific
AC441181000
100mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441185000 SDS Thermo Fisher Scientific
AC441185000
500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

121786-40-1
100.0
C7H11NO3
(2R)-2-(Acetylamino)-4-pentenoic acid
≥97.5%
97.5
98%
157.17
(R)-N-Acetyl-α-allylglycine, 98% ee

Specifications

121786-40-1
100.0
Authentic
98%
C7H11NO3
(2R)-2-(Acetylamino)-4-pentenoic acid
100mg
≥97.5%
97.5
98.0% min.
Powder
-6° to -10° (20°C, 589nm) (c=1 in water)
157.17
White to pale yellow
(R)-N-Acetyl-α-allylglycine, 98% ee