Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(+)-N-(3-Pentyl)-1-phenylethylamine hydrochloride, 99%

$216.41 - $216.41

Chemical Identifiers

Chemical Name or Material (R)-(+)-N-(3-Pentyl)-1-phenylethylamine hydrochloride
CAS 374790-91-7
Molecular Formula C13H21N·HCl
Formula Weight 227.78g/mol
MDL Number MFCD06199156
Synonym (R)-N-(1-Ethylpropyl)-1-phenylethylamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1835606

 
alfa aesar™
L18356-06
5g Each for $216.41
Description & Specifications

Specifications

Melting Point 232° to 234°C
Quantity 5g
Optical Rotation +17° (c=1 in Water)
Chemical Name or Material (R)-(+)-N-(3-Pentyl)-1-phenylethylamine hydrochloride
CAS 374790-91-7
Assay 99%
Molecular Formula C13H21N·HCl
Formula Weight 227.78g/mol
MDL Number MFCD06199156
Synonym (R)-N-(1-Ethylpropyl)-1-phenylethylamine hydrochloride
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.