Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine

NMR chiral shift reagent

Chemical Identifiers

Chemical Name or Material (R)-(-)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
CAS 69632-32-2
Formula Weight 315.28
MDL Number MFCD00010866
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2268402

  SDS
alfa aesar™
2268402
0.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA2268477

  SDS
alfa aesar™
2268477
0.1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 315.28
Optical Rotation −48°C (c=1 in Acetone)
Chemical Name or Material (R)-(-)-N-(3,5-Dinitrobenzoyl)-1-phenylethylamine
Physical Form Crystalline
MDL Number MFCD00010866
CAS 69632-32-2
Color Off-white to pale yellow
Quantity 0.5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 158° to 160°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.