missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-α-Methyltyrosine, 98%, 98% ee, ACROS Organics™

Manufacturer:  Acros Organics AC441242500

 View more versions of this product

Catalog No. AC441242500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(R)-α-Methyltyrosine
Authentic
White to Yellow
98% ee
97.5
98%
r-alpha-methyltyrosine, alpha-methyl-d-tyr, r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, 2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, alpha-methyl-d-tyrosine, metirosinum inn-latin, metirosina inn-spanish, l-tyrosine, alpha-methyl-,-, d +-metyrosine, metyrosine usan
CC(CC1=CC=C(C=C1)O)(C(=O)O)N
195.218
195.22
≥97.5%
98.0% min.
+15° to +21° (20°C, 589nm) (c=0.05 in water)
250mg
672-86-6
100.0
C10H13NO3
NHTGHBARYWONDQ-SNVBAGLBSA-N
(2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
12650
Powder

Chemical Identifiers

672-86-6
195.218
r-alpha-methyltyrosine, alpha-methyl-d-tyr, r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, 2r-2-amino-3-4-hydroxyphenyl-2-methylpropanoic acid, alpha-methyl-d-tyrosine, metirosinum inn-latin, metirosina inn-spanish, l-tyrosine, alpha-methyl-,-, d +-metyrosine, metyrosine usan
(2R)-2-amino-3-(4-hydroxyphenyl)-2-methylpropanoic acid
C10H13NO3
NHTGHBARYWONDQ-SNVBAGLBSA-N
12650
CC(CC1=CC=C(C=C1)O)(C(=O)O)N
SDS Documents