Please login to your online account to display your discounted pricing

(R)-α-Methyltyrosine, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

Chemical Name or Material (R)-α-Methyltyrosine
Name Note 98% ee
CAS 672-86-6
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H13NO3
Formula Weight 195.22
Synonym (R)-α-Methyl-4-hydroxyphenylalanine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC441240010

  SDS
thermo fisher scientific
AC441240010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC441242500

  SDS
thermo fisher scientific
AC441242500
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder
Assay ≥97.5%
Formula Weight 195.22
Quantity 1g
Color White to pale yellow
CAS Max % 100.0
Chemical Name or Material (R)-α-Methyltyrosine
Name Note 98% ee
Assay Percent Range 98%
Molecular Formula C10H13NO3
CAS 672-86-6
CAS Min % 97.5
Infrared Spectrum Authentic
Specific Rotation +15° to +21° (20°C, 589nm) (c=0.05 in water)
Synonym (R)-α-Methyl-4-hydroxyphenylalanine
Enantiomeric Excess 98.0% min.