missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-α-Methylphenylalanine hydrate, 98%, 98% ee, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC441291000

 View more versions of this product

Catalog No. AC441291000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

(R)-α-Methylphenylalanine hydrate
Authentic
White to Yellow
100mg
17350-84-4
100.0
C10H13NO2·xH2O
HYOWVAAEQCNGLE-SNVBAGLBSA-N
(2R)-2-amino-2-methyl-3-phenylpropanoic acid
7023409
Powder
98.0% min.
18
>233°C
98% ee
97.5
97.5% min. (HPLC)
2r-2-amino-2-methyl-3-phenylpropanoic acid, a-benzyl-d-ala, alpha-benzyl-d-ala, alpha-methyl-d-phe, alpha-methyl-d-phenylalanine, d-alpha-methyl phenylalanine, h-alpha-me-d-phe-oh, r-2-amimo-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropionic acid
CC(CC1=CC=CC=C1)(C(=O)O)N
197.23
197.23
98%,98% ee

Chemical Identifiers

17350-84-4
197.23
2r-2-amino-2-methyl-3-phenylpropanoic acid, a-benzyl-d-ala, alpha-benzyl-d-ala, alpha-methyl-d-phe, alpha-methyl-d-phenylalanine, d-alpha-methyl phenylalanine, h-alpha-me-d-phe-oh, r-2-amimo-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropionic acid
(2R)-2-amino-2-methyl-3-phenylpropanoic acid
C10H13NO2·xH2O
HYOWVAAEQCNGLE-SNVBAGLBSA-N
7023409
CC(CC1=CC=CC=C1)(C(=O)O)N
SDS Documents