missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-α-Methylphenylalanine hydrate, 98%, 98% ee, ACROS Organics™

Chemical Identifiers

CAS 17350-84-4
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C10H13NO2·xH2O
Formula Weight 179.22
Synonym (R)-2-Amino-2-methyl-3-phenylpropionic acid hydrate
Chemical Name or Material (R)-α-Methylphenylalanine hydrate, 98% ee
Assay ≥97.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC441291000 SDS Thermo Fisher Scientific
AC441291000
100mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC441295000 SDS Thermo Fisher Scientific
AC441295000
500mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

17350-84-4
100.0
C10H13NO2·xH2O
(R)-2-Amino-2-methyl-3-phenylpropionic acid hydrate
≥97.5%
97.5
98%
179.22
(R)-α-Methylphenylalanine hydrate, 98% ee

Specifications

Powder
100mg
100.0
C10H13NO2·xH2O
97.5
(R)-2-Amino-2-methyl-3-phenylpropionic acid hydrate
Authentic
≥97.5%
179.22
White to pale yellow
+16° to +23° (20°C, 589nm) (c=1 in H2O)
17350-84-4
98%
98.0% min.
(R)-α-Methylphenylalanine hydrate, 98% ee