missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-α-Methylphenylalanine hydrate, 98%, 98% ee, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441295000
Specifications
(R)-α-Methylphenylalanine hydrate | |
Authentic | |
White to Yellow | |
500mg | |
17350-84-4 | |
100.0 | |
C10H13NO2·xH2O | |
HYOWVAAEQCNGLE-SNVBAGLBSA-N | |
(2R)-2-amino-2-methyl-3-phenylpropanoic acid | |
7023409 | |
Powder |
98.0% min. | |
18 | |
>233°C | |
98% ee | |
97.5 | |
97.5% min. (HPLC) | |
2r-2-amino-2-methyl-3-phenylpropanoic acid, a-benzyl-d-ala, alpha-benzyl-d-ala, alpha-methyl-d-phe, alpha-methyl-d-phenylalanine, d-alpha-methyl phenylalanine, h-alpha-me-d-phe-oh, r-2-amimo-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropionic acid | |
CC(CC1=CC=CC=C1)(C(=O)O)N | |
197.23 | |
197.23 | |
98%,98% ee |
Chemical Identifiers
17350-84-4 | |
197.23 | |
2r-2-amino-2-methyl-3-phenylpropanoic acid, a-benzyl-d-ala, alpha-benzyl-d-ala, alpha-methyl-d-phe, alpha-methyl-d-phenylalanine, d-alpha-methyl phenylalanine, h-alpha-me-d-phe-oh, r-2-amimo-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropanoic acid, r-2-amino-2-methyl-3-phenylpropionic acid | |
(2R)-2-amino-2-methyl-3-phenylpropanoic acid |
C10H13NO2·xH2O | |
HYOWVAAEQCNGLE-SNVBAGLBSA-N | |
7023409 | |
CC(CC1=CC=CC=C1)(C(=O)O)N |