Please login to your online account to display your discounted pricing

(R)-(-)-α-Methylhistamine dihydrobromide, Tocris

Potent, standard H3 agonist

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material (R)-(-)-α-Methylhistamine dihydrobromide
Assay Percent Range >99%
Molecular Formula C6H11N3·2HBr
Formula Weight 287
Synonym (R)-(-)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

056910

 
r & d systems
RDS056910
10mg Each for $168.15

056950

 
r & d systems
RDS056950
50mg Each for $697.40
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 287
Chemical Name or Material (R)-(-)-α-Methylhistamine dihydrobromide
Assay Percent Range >99%
Molecular Formula C6H11N3·2HBr
CAS 868698-49-1
Synonym (R)-(-)-α-Methyl-1H-imidazole-4-ethanamine dihydrobromide
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature