missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-alpha-Methylaspartic acid, 98%, 98% ee, ACROS Organics™
Chemical Identifiers
CAS | 14603-76-0 |
---|---|
Molecular Formula | C5H9NO4 |
Molecular Weight (g/mol) | 147.13 |
InChI Key | CWAYDJFPMMUKOI-RXMQYKEDSA-N |
Synonym | 2-methyl-d-aspartic acid, 2r-2-amino-2-methylbutanedioic acid, a-methyl-d-asp, alpha-methyl-d-asp, ambotzhaa7540, d-aspartic acid,2-methyl, d-aspartic acid,2-methyl-, hydrochloride 1:1, h-a-me-d-asp-oh, r---2-amino-2-methylbutanedioic acid, r-2-methylaspartic acid |
PubChem CID | 6993662 |
IUPAC Name | (2R)-2-amino-2-methylbutanedioic acid |
SMILES | CC(CC(=O)O)(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC290141000
|
Thermo Fisher Scientific
AC290141000 |
100mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC290145000
|
Thermo Fisher Scientific
AC290145000 |
500mg |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
14603-76-0 | |
147.13 | |
2-methyl-d-aspartic acid, 2r-2-amino-2-methylbutanedioic acid, a-methyl-d-asp, alpha-methyl-d-asp, ambotzhaa7540, d-aspartic acid,2-methyl, d-aspartic acid,2-methyl-, hydrochloride 1:1, h-a-me-d-asp-oh, r---2-amino-2-methylbutanedioic acid, r-2-methylaspartic acid | |
(2R)-2-amino-2-methylbutanedioic acid |
C5H9NO4 | |
CWAYDJFPMMUKOI-RXMQYKEDSA-N | |
6993662 | |
CC(CC(=O)O)(C(=O)O)N |
Specifications
> 75% | |
-54 | |
>249°C | |
14603-76-0 | |
100.0 | |
C5H9NO4 | |
2-methyl-d-aspartic acid, 2r-2-amino-2-methylbutanedioic acid, a-methyl-d-asp, alpha-methyl-d-asp, ambotzhaa7540, d-aspartic acid,2-methyl, d-aspartic acid,2-methyl-, hydrochloride 1:1, h-a-me-d-asp-oh, r---2-amino-2-methylbutanedioic acid, r-2-methylaspartic acid | |
CC(CC(=O)O)(C(=O)O)N | |
147.13 | |
147.13 | |
98%,98% ee |
Authentic | |
White | |
100mg | |
97.5 | |
97.5% min | |
HOOCC(CH3)NH2CH2COOH | |
CWAYDJFPMMUKOI-RXMQYKEDSA-N | |
(2R)-2-amino-2-methylbutanedioic acid | |
6993662 | |
Fine Crystalline Powder | |
(R)-α-Methylaspartic acid, 99+%, (80% e.e.) |
Safety and Handling
TSCA : TSCA