missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(-)-alpha-Methoxyphenylacetic acid, 99%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 224470050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chiral derivatization reagentSpecifications
(R)-(-)-α-Methoxyphenylacetic acid | |
98.5 | |
Authentic | |
Glass bottle | |
C6H5CH(OCH3)COOH | |
5g | |
−160.40° (20°C c=1,methanol) | |
r---alpha-methoxyphenylacetic acid, r-2-methoxy-2-phenylacetic acid, 2r-2-methoxy-2-phenylacetic acid, r-methoxyphenylacetic acid, r-methoxy phenyl acetic acid, methoxy phenyl acetic acid #, r---.alpha.-methoxyphenylacetic acid, benzeneacetic acid, .alpha.-methoxy-, r, o-methyl-d-mandelic acid, ksc496q9j | |
COC(C1=CC=CC=C1)C(=O)O | |
166.18 | |
166.18 |
3966-32-3 | |
100.0 | |
99% | |
C9H10O3 | |
MFCD00004250 | |
10, 195 | |
-160.4 | |
DIWVBIXQCNRCFE-MRVPVSSYSA-N | |
(2R)-2-methoxy-2-phenylacetic acid | |
2724294 | |
99% |
Safety and Handling
EINECSNumber : 223-580-9
RUO â Research Use Only