Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-Mandelic acid, ChiPros 99+%, ee 99+%

$30.93 - $97.28

Chemical Identifiers

Chemical Name or Material (R)-(-)-Mandelic acid
CAS 611-71-2
Molecular Formula C8H8O3
Formula Weight 152.15
MDL Number MFCD00064251
Beilstein 2691094
Synonym (R)-(-)-alpha-Hydroxyphenylacetic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1911606

 
alfa aesar™
L1911606
5g Each for $30.93

AAL1911614

 
alfa aesar™
L1911614
25g Each for $97.28
Description & Specifications

Specifications

Formula Weight 152.15
Assay ≥99%
Molecular Formula C8H8O3
Beilstein 2691094
Melting Point 131° to 135°C
Synonym (R)-(-)-alpha-Hydroxyphenylacetic acid
Optical Rotation −153° (c=5 in Water)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material (R)-(-)-Mandelic acid
TSCA TSCA
Assay Percent Notes ee ≥99%
MDL Number MFCD00064251
Health Hazard 2 GHS H Statement
H318
Causes serious eye damage.
CAS 611-71-2
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H318
Causes serious eye damage.