Learn More
(R)-(-)-Mandelic acid, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1476714
Description
(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.
Solubility
It is partly soluble in water, freely soluble in isopropyl and ethyl alcohol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under ordinary conditions. Incompatabile to strong bases, strong oxidizing agents.
Specifications
(R)-(-)-Mandelic acid | |
611-71-2 | |
MFCD00064251 | |
It is partly soluble in water,freely soluble in isopropyl and ethyl alcohol. | |
O[C@@H](C(O)=O)C1=CC=CC=C1 | |
152.15 | |
CHEBI:17656 | |
98% | |
-153° (c=5 in Water) |
130°C to 134°C | |
C8H8O3 | |
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid | |
IWYDHOAUDWTVEP-SSDOTTSWSA-N | |
(2R)-2-hydroxy-2-phenylacetic acid | |
11914 | |
152.15 | |
25 g | |
Light sensitive |
Chemical Identifiers
611-71-2 | |
152.15 | |
IWYDHOAUDWTVEP-SSDOTTSWSA-N | |
11914 | |
(2R)-2-hydroxy-2-phenylacetic acid |
C8H8O3 | |
MFCD00064251 | |
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, #NAME?, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid | |
CHEBI:17656 | |
O[C@@H](C(O)=O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
EINECSNumber : 210-276-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only