missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-Mandelic acid, 98%

Manufacturer:  Alfa Aesar™ A1476706

 View more versions of this product

Catalog No. AAA1476706



Description

Description

Applications
(R)-(-)-Mandelic acid, is used as a antiseptic ingredient particularly against urinary tract infections.Mandelic acid and its derivatives are used to apply the dual activities as an antibacterial agent and as an antiaging agent. It is used as an intermediate for the synthesis of target molecules for other applications.

Solubility
It is partly soluble in water, freely soluble in isopropyl and ethyl alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Light sensitive. Stable under ordinary conditions. Incompatabile to strong bases, strong oxidizing agents.
Specifications

Specifications

(R)-(-)-Mandelic acid
130°C to 134°C
611-71-2
MFCD00064251
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
C1=CC=C(C=C1)C(C(=O)O)O
152.149
CHEBI:17656
98%
−153° (c=5 in Water)
5g
C8H8O3
2691094
IWYDHOAUDWTVEP-SSDOTTSWSA-N
(2R)-2-hydroxy-2-phenylacetic acid
11914
152.15

Chemical Identifiers

611-71-2
152.149
IWYDHOAUDWTVEP-SSDOTTSWSA-N
11914
(2R)-2-hydroxy-2-phenylacetic acid
C8H8O3
MFCD00064251
r-mandelic acid, r---mandelic acid, d-mandelic acid, r-2-hydroxy-2-phenylacetic acid, --mandelic acid, d--mandelic acid, d---mandelic acid, 2r-2-hydroxy-2-phenylacetic acid, r-alpha-hydroxyphenylacetic acid, d-2-phenylglycolic acid
CHEBI:17656
C1=CC=C(C=C1)C(C(=O)O)O
Safety and Handling

Safety and Handling

GHS H Statement
H318
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

Danger

TSCA : TSCA

SDS Documents