missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-(+)-Lactamide 98.0+%, TCI America™
Supplier: TCI America L016725G
Specifications
(R)-(+)-Lactamide | |
54°C | |
MFCD00210310 | |
(R)-(+)-2-Hydroxypropionamide | |
CC(C(=O)N)O | |
89.094 | |
89.09 | |
Crystalline Powder |
598-81-2 | |
C3H7NO2 | |
25 g | |
SXQFCVDSOLSHOQ-UWTATZPHSA-N | |
(2R)-2-hydroxypropanamide | |
1530236 | |
≥98.0% (GC,N) |
Chemical Identifiers
598-81-2 | |
89.094 | |
SXQFCVDSOLSHOQ-UWTATZPHSA-N | |
1530236 | |
CC(C(=O)N)O |
C3H7NO2 | |
MFCD00210310 | |
(R)-(+)-2-Hydroxypropionamide | |
(2R)-2-hydroxypropanamide |
Safety and Handling
TSCA : No