missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(+)-HA-966, Tocris

NMDA partial agonist/antagonist, acts at glycine site

$181.19 - $181.19

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C4H8N2O2
Formula Weight 116.12
Synonym (R)-(+)-3-Amino-1-hydroxypyrrolidin-2-one
Chemical Name or Material (R)-(+)-HA-966
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
028110 R & D Systems
RDS028110
10mg Each for $181.19
Specifications Description & Specifications

Chemical Identifiers

>98%
116.12
(R)-(+)-HA-966
C4H8N2O2
(R)-(+)-3-Amino-1-hydroxypyrrolidin-2-one

Specifications

123931-04-4
C4H8N2O2
10mg
(R)-(+)-HA-966
>98%
116.12
(R)-(+)-3-Amino-1-hydroxypyrrolidin-2-one

Safety and Handling

Recommended Storage : Desiccate at Room Temperature