missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(R)-Glycidyl Butyrate 98.0+%, TCI America™
$90.69 - $272.06
Chemical Identifiers
CAS | 60456-26-0 |
---|---|
Molecular Formula | C7H12O3 |
Molecular Weight (g/mol) | 144.17 |
MDL Number | MFCD00075120 |
InChI Key | YLNSNVGRSIOCEU-ZCFIWIBFSA-N |
Synonym | 2r oxiran-2-yl methyl butanoate, 2r-oxiran-2-yl methyl butanoate, 2r-oxiran-2-ylmethyl butanoate, butanoic acid, 2r-oxiranylmethyl ester, butyric acid r-glycidyl ester, r---glycidyl butyrate, r-glycicyl butanoate, r-glycidyl butyrate, r-oxiran-2-ylmethyl butyrate |
PubChem CID | 2724536 |
IUPAC Name | [(2R)-oxiran-2-yl]methyl butanoate |
SMILES | CCCC(=O)OCC1CO1 |
Chemical Identifiers
60456-26-0 | |
144.17 | |
YLNSNVGRSIOCEU-ZCFIWIBFSA-N | |
2724536 | |
CCCC(=O)OCC1CO1 |
C7H12O3 | |
MFCD00075120 | |
2r oxiran-2-yl methyl butanoate, 2r-oxiran-2-yl methyl butanoate, 2r-oxiran-2-ylmethyl butanoate, butanoic acid, 2r-oxiranylmethyl ester, butyric acid r-glycidyl ester, r---glycidyl butyrate, r-glycicyl butanoate, r-glycidyl butyrate, r-oxiran-2-ylmethyl butyrate | |
[(2R)-oxiran-2-yl]methyl butanoate |
Specifications
90°C | |
5g | |
C7H12O3 | |
2r oxiran-2-yl methyl butanoate, 2r-oxiran-2-yl methyl butanoate, 2r-oxiran-2-ylmethyl butanoate, butanoic acid, 2r-oxiranylmethyl ester, butyric acid r-glycidyl ester, r---glycidyl butyrate, r-glycicyl butanoate, r-glycidyl butyrate, r-oxiran-2-ylmethyl butyrate | |
CCCC(=O)OCC1CO1 | |
144.17 | |
144.17 | |
≥98.0% (GC) |
Yellow | |
60456-26-0 | |
MFCD00075120 | |
YLNSNVGRSIOCEU-ZCFIWIBFSA-N | |
[(2R)-oxiran-2-yl]methyl butanoate | |
2724536 | |
Clear Liquid at 20°C | |
(R)-Glycidyl Butyrate |
Safety and Handling
TSCA : No