Please login to your online account to display your discounted pricing

Alfa Aesar™ (R)-(-)-Glycidyl butyrate, 98%

$38.95 - $120.84

Chemical Identifiers

Chemical Name or Material (R)-(-)-Glycidyl butyrate
CAS 60456-26-0
Molecular Formula C7H12O3
Formula Weight 144.17
MDL Number MFCD00075120
Beilstein 5246881
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1855703

  SDS
alfa aesar™
L1855703
1g Each for $38.95

AAL1855706

  SDS
alfa aesar™
L1855706
5g Each for $120.84

AAL1855714

  SDS
alfa aesar™
L1855714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 144.17
Chemical Name or Material (R)-(-)-Glycidyl butyrate
Assay 98%
MDL Number MFCD00075120
CAS 60456-26-0
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Molecular Formula C7H12O3
Beilstein 5246881
Health Hazard 1 Warning
Boiling Point 90°C (19mmHg)
Flash Point 85°C (185°F)
Optical Rotation −31° (Neat)
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H315-H319-H335-H227
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.