missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-Glycidyl butyrate, 92-94%, ACROS Organics™

$168.67 - $168.67

Chemical Identifiers

CAS 60456-26-0
CAS Min % 92.0
CAS Max % 100.0
Assay Percent Range 92-94%
Molecular Formula C7H12O3
Formula Weight 144.17
MDL Number MFCD00075120
Chemical Name or Material (R)-Glycidyl butyrate, 92-94%
Assay ≥92%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC252830100 SDS Thermo Fisher Scientific
252830100
Glass bottle 10g Each for $168.67
Specifications Description & Specifications

Chemical Identifiers

60456-26-0
100.0
C7H12O3
MFCD00075120
≥92%
92.0
92-94%
144.17
(R)-Glycidyl butyrate, 92-94%

Specifications

60456-26-0
100.0
Liquid
Glass bottle
144.17
1.4270 to 1.4290 (20°C, 589nm)
1.0350g/mL
Clear colorless to slightly yellow
(R)-Glycidyl butyrate, 92-94%
92.0
Authentic
92-94%
C7H12O3
MFCD00075120
-29° to -32° (20°C, 589nm) (neat)
− 30.00 (20.00°C c=neat)
10g
≥92%