Please login to your online account to display your discounted pricing

(R)-(+)-Etomoxir sodium salt, Tocris™

Carnitine palmitoyltransferase I (CPT1) inhibitor

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material (R)-(+)-Etomoxir sodium salt
Assay Percent Range >98%
Molecular Formula C15H18ClNaO4
Formula Weight 320.74
Synonym (R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

453910

 
r & d systems
RDS453910
10mg Each for $214.12

453950

 
r & d systems
RDS453950
50mg Each for $895.76
Description & Specifications

Specifications

Formula Weight 320.74
Quantity 10mg
Synonym (R)-(+)-2-[6-(4-Chlorophenoxy)hexyl]-oxirane-2-carboxylic acid sodium salt
CAS 828934-41-4
Chemical Name or Material (R)-(+)-Etomoxir sodium salt
Assay Percent Range >98%
Molecular Formula C15H18ClNaO4
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C