missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(R)-(-)-Deprenyl hydrochloride, Thermo Scientific Chemicals

Selective MAO-B inhibitor; anti-Parkinsonian agent

Supplier:  Thermo Scientific Chemicals J61286MD

 View more versions of this product

Catalog No. AAJ61286MD



Description

Description

(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-Deprenyl hydrochloride provides neuroprotection which may be relevant to Parkinson’s disease, Alzheimer’s disease, Huntington’s disease, and stroke. It is an inhibitor of MAO-B (monoamine oxidase-B) which demonstrates extensive neuroprotective effects shown to be uncorrelated with MAO-B inhibiting activity. It demonstrates protection of dopaminergic neurons from excitotoxicity mediated by NMDA activation. It also demonstrates decreases in apoptosis of PC12 neuronal cells accompanied with an induction of new protein synthesis through direct transcriptional and translational interaction, and reduced cell death and internucleosomal DNA degredation at concentrations too low to inhibit MAO-B.

Solubility
Soluble in water at 100mM

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

(R)-(-)-Deprenyl hydrochloride
White
MFCD00069299
14,8428
Soluble in water at 100mM
−12° (c=1 in Water)
hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
26758
223.74
14611-52-0
C13H18ClN
250 mg
selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
IYETZZCWLLUHIJ-UTONKHPSSA-N
[H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C
223.74
CHEBI:9087
Crystalline Powder

Chemical Identifiers

14611-52-0
223.74
IYETZZCWLLUHIJ-UTONKHPSSA-N
26758
hydrogen methyl[(2R)-1-phenylpropan-2-yl](prop-2-yn-1-yl)amine chloride
C13H18ClN
MFCD00069299
selegiline hydrochloride, eldepryl, zelapar, selegiline hcl, l-deprenyl hydrochloride, jumex hydrochloride, --deprenyl hydrochloride, eldepryl hydrochloride, r-n-methyl-n-1-phenylpropan-2-yl prop-2-yn-1-amine hydrochloride, zydis selegiline
CHEBI:9087
[H+].[Cl-].C[C@H](CC1=CC=CC=C1)N(C)CC#C
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

RTECSNumber : DA0292500

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only