missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Zebrafish BMP-4 Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 30612
Name BMP-4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Price Quantity  
1128BM010 R&D Systems
RDS1128BM010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. 12.9 kDa Each for $416.79
1128BM010CF R&D Systems
RDS1128BM010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. 12.9 kDa (monomer) Each for $416.79
Specifications

Description

Source: E. coli-derived, Ser288-Arg400, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 12.9 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Entrez Gene ID: 30612
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
BMP-4
10μg
Zebrafish
30612
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes