missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Viral MIP-II Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Molecular Weight 8.1 kDa
Name MIP-II
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
601VB025CF R&D Systems
RDS601VB025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $428.87
601VB025 R&D Systems
RDS601VB025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $428.87
Specifications

Description

Source: E. coli-derived, Leu24-Arg94
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 8.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
MIP-II
25μg
Viral
8.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes