missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Viral MCV-type II Chemokine-like Protein Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Molecular Weight 9.1 kDa
Name MCV-type II Chemokine-like Protein
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
697M3025 R&D Systems
RDS697M3025
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $376.73
697M3025CF R&D Systems
RDS697M3025CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $376.73
Specifications

Description

Source: E. coli-derived, Leu25-Leu104
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 9.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
MCV-type II Chemokine-like Protein
25μg
Viral
9.1 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes