Please login to your online account to display your discounted pricing

R&D Systems Viral HCMV IL-10 Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2949786
Molecular Weight 17.6 kDa
Name HCMV IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

117VL025

 
r&d systems
RDS117VL025
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. Each for $419.81

117VL025CF

 
r&d systems
RDS117VL025CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2949786
Molecular Weight 17.6 kDa
Name HCMV IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Viral

Source: E. coli-derived, Ala26-Lys176
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.6 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 2949786
Regulatory Status: RUO