missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Viral EBV IL-10 Recombinant Protein

$428.87 - $428.87

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 2949786
Molecular Weight 17.2 kDa
Name EBV IL-10
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
915VL010 R&D Systems
RDS915VL010
Lyophilized from a 0.2μm filtered solution in Tris and NaCl with BSA as a carrier protein. Each for $428.87
915VL010CF R&D Systems
RDS915VL010CF
Supplied as a 0.2μm filtered solution in Tris and NaCl. Each for $428.87
Specifications

Description

Source: E. coli-derived, Gln26-Arg170, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.2 kDa
Formulation: Supplied as a 0.2μm filtered solution in Tris and NaCl.
Entrez Gene ID: 2949786
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
2949786
EBV IL-10
10μg
Viral