Please login to your online account to display your discounted pricing

R&D Systems Viral CCI Fc Chimera (CHO-expressed) Recombinant Protein

$419.81 - $419.81

Specifications

Conjugate Unlabeled
Molecular Weight 52.6 kDa (monomer)
Name CCI Fc Chimera (CHO-expressed)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

7055CC025

 
r&d systems
RDS7055CC025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $419.81

7055CC025CF

 
r&d systems
RDS7055CC025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $419.81
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 52.6 kDa (monomer)
Name CCI Fc Chimera (CHO-expressed)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Viral

Source: Chinese Hamster Ovary cell line, CHO-derived, Met1-Val258
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 52.6 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO