missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rhesus Macaque TNF-alpha Recombinant Protein

$272.44 - $272.44

Specifications

Conjugate Unlabeled
Molecular Weight 17.4 kDa
Name TNF-alpha
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1070RM025CF R&D Systems
RDS1070RM025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $272.44
1070RM025 R&D Systems
RDS1070RM025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. Each for $272.44
Specifications

Description

Source: E. coli-derived, Val77-Leu233
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
TNF-alpha
25μg
Rhesus Macaque
17.4 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes