missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rhesus Macaque IL-18/IL-1F4 Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Molecular Weight 18.2 kDa
Name IL-18/IL-1F4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
2548RM025 R&D Systems
RDS2548RM025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $376.73
2548RM025CF R&D Systems
RDS2548RM025CF
Supplied as a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $376.73
Specifications

Description

Source: E. coli-derived, Tyr37-Asp193
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.2 kDa
Formulation: Supplied as a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-18/IL-1F4
25μg
Rhesus Macaque
18.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes