Please login to your online account to display your discounted pricing

R&D Systems Rhesus Macaque IL-1 beta/IL-1F2 Recombinant Protein


Manufacturer: r&d systems  RDS1318RL010CF

 View more versions of this product

Catalog No. 1318RL010CF

  • / Each
 

Description & Specifications

Specifications

Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Quantity 10μg
Molecular Weight 17.3 kDa
Name IL-1 beta/IL-1F2
Recombinant Yes
Source Rhesus Macaque
Conjugate Unlabeled