missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rhesus Macaque IFN-gamma Recombinant Protein

$376.73 - $376.73

Specifications

Conjugate Unlabeled
Molecular Weight 16.9 kDa
Name IFN-gamma
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
961RM025CF R&D Systems
RDS961RM025CF
Lyophilized from a 0.2μm filtered solution in Tris. Each for $376.73
961RM025 R&D Systems
RDS961RM025
Lyophilized from a 0.2μm filtered solution in Tris with BSA as a carrier protein. Each for $376.73
Specifications

Description

Source: E. coli-derived, Gln24-Gln165, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.9 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Tris.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IFN-gamma
25μg
Rhesus Macaque
16.9 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes