Please login to your online account to display your discounted pricing

R&D Systems Recombinant Mouse IFN-β

Interferon beta

$311.55 - $311.55

Specifications

Quantity 10μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

82-34M-B010

 
r&d systems
8234MB010
Lyophilized from a 0.2μm filtered solution in PBS and Tween™ 80 with BSA as a carrier protein Each for $311.55
Description & Specifications

Specifications

Quantity 10μg

Interferon-beta (IFN-beta), along with IFN-alpha and IFN-omega, is a member of the type I interferon family. All three type I interferons share a common cell surface receptor that is composed of two subunits, a 100 kDa ligand-binding subunit (IFN-alpha R2) and a 125 kDa subunit (IFN-alpha R1) that is involved both in ligand binding and signal transduction. Type I interferons were originally described as potent anti-viral molecules produced by virally infected cells.

Source: Human embryonic kidney cell, HEK293-derived, Ile22-Asn182
Accession No.: P01575
N-terminal Sequence Analysis: Ile22
Structure: Monomer
Purity: >95%, by SDS-PAGE with silver staining
Predicted Molecular Mass: 20 kDa
SDS-PAGE: 30-38 kDa, reducing conditions
Activity: Measured in an anti-viral assay using LG–929 mouse fibroblast cells infected with encephalomyocarditis (EMC) virus

The product is shipped at ambient temperature, upon receipt, store immediately