missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat VEGF 164 Recombinant Protein

$415.83 - $1,184.94

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 83785
Molecular Weight 19.2 kDa (monomer)
Name VEGF 164
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
564RV010 R&D Systems
RDS564RV010
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10μg Each for $415.83
564RV010CF R&D Systems
RDS564RV010CF
Lyophilized from a 0.2μm filtered solution in PBS. 10μg Each for $415.83
564RV050CF R&D Systems
RDS564RV050CF
Lyophilized from a 0.2μm filtered solution in PBS. 50μg Each for $1,184.94
564RV050 R&D Systems
RDS564RV050
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50μg Each for $1,184.94
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Ala27-Arg190
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19.2 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 83785
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
19.2 kDa (monomer)
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
83785
VEGF 164
Yes