missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat TNF-alpha Recombinant Protein

$298.51 - $911.19

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24835
Molecular Weight 17 kDa
Name TNF-alpha
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
510RT010 R&D Systems
RDS510RT010
Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein. 10μg Each for $298.51
510RT010CF R&D Systems
RDS510RT010CF
Lyophilized from a 0.2μm filtered solution in PBS and DTT. 10μg Each for $298.51
510RT050 R&D Systems
RDS510RT050
Lyophilized from a 0.2μm filtered solution in PBS and DTT with BSA as a carrier protein. 50μg Each for $911.19
510RT050CF R&D Systems
RDS510RT050CF
Lyophilized from a 0.2μm filtered solution in PBS and DTT. 50μg Each for $911.19
Specifications

Description

Source: E. coli-derived, Leu80-Leu235, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS and DTT.
Entrez Gene ID: 24835
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
24835
TNF-alpha
Yes