missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat Pentraxin 2/SAP Recombinant Protein

$416.79 - $416.79

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 29339
Molecular Weight 25 kDa
Name Pentraxin 2/SAP
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1895SA050CF R&D Systems
RDS1895SA050CF
Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CaCl2. Each for $416.79
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln21-Ser228 with a C-terminal 10-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 25 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, NaCl and CaCl2.
Entrez Gene ID: 29339
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
25 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
29339
Pentraxin 2/SAP
50μg
Rat