missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat Leptin/OB Recombinant Protein

$272.44 - $676.55

Specifications

Conjugate Unlabeled
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 25608
Molecular Weight 16.2 kDa
Name Leptin/OB
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
598LP01M R&D Systems
RDS598LP01M
1mg Each for $272.44
598LP05M R&D Systems
RDS598LP05M
5mg Each for $676.55
Specifications

Description

Source: E. coli-derived, Val22-Cys167, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25608
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
25608
Leptin/OB
Yes
Lyophilized from a 0.2μm filtered solution in PBS.
16.2 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat