missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-9 Recombinant Protein

$298.51 - $298.51

Specifications

Conjugate Unlabeled
Molecular Weight 14.1 kDa
Name IL-9
Purity 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
4134RL010 R&D Systems
RDS4134RL010
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. Each for $298.51
4134RL010CF R&D Systems
RDS4134RL010CF
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. Each for $298.51
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Gln19-Ala144
Purity: 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.1 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA.
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-9
10μg
Rat
14.1 kDa
90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes