missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-6 Recombinant Protein

$272.44 - $846.01

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24498
Molecular Weight 22 kDa
Name IL-6
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
506RL010CF R&D Systems
RDS506RL010CF
Supplied as a 0.2μm filtered solution in Sodium Acetate. 10μg Each for $272.44
506RL010 R&D Systems
RDS506RL010
Supplied as a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. 10μg Each for $272.44
506RL050 R&D Systems
RDS506RL050
Supplied as a 0.2μm filtered solution in Sodium Acetate with BSA as a carrier protein. 50μg Each for $846.01
506RL050CF R&D Systems
RDS506RL050CF
Supplied as a 0.2μm filtered solution in Sodium Acetate. 50μg Each for $846.01
Specifications

Description

Source: E. coli-derived, Phe25-Thr211, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 22 kDa
Formulation: Supplied as a 0.2μm filtered solution in Sodium Acetate.
Entrez Gene ID: 24498
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
22 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
24498
IL-6
Yes