missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-4 Recombinant Protein

$272.44 - $814.73

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 287287
Molecular Weight 13.5 kDa
Name IL-4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
504RL025 R&D Systems
RDS504RL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $814.73
504RL005CF R&D Systems
RDS504RL005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $272.44
504RL005 R&D Systems
RDS504RL005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $272.44
504RL025CF R&D Systems
RDS504RL025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $814.73
Specifications

Description

Source: E. coli-derived, Cys25-Ser147, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 13.5 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 287287
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
13.5 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
287287
IL-4
Yes