missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-2 Recombinant Protein

$311.55 - $997.23

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 116562
Molecular Weight 16 kDa
Name IL-2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
502RL050 R&D Systems
RDS502RL050
Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol with BSA as a carrier protein. 50μg Each for $997.23
502RL010CF R&D Systems
RDS502RL010CF
Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol. 10μg Each for $311.55
502RL010 R&D Systems
RDS502RL010
Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol with BSA as a carrier protein. 10μg Each for $311.55
502RL050CF R&D Systems
RDS502RL050CF
Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol. 50μg Each for $997.23
Specifications

Description

Source: E. coli-derived, Ala21-Gln155, with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16 kDa
Formulation: Supplied as a 0.2μm filtered solution in Ammonium Acetate and Glycerol.
Entrez Gene ID: 116562
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
16 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
116562
IL-2
Yes