missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-18/IL-1F4 Recombinant Protein

$290.11 - $290.11

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 29197
Molecular Weight 18.4 kDa
Name IL-18/IL-1F4
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
521RL025 R&D Systems
RDS521RL025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA, DTT and trehalose with BSA as a carrier protein. Each for $290.11
521RL025CF R&D Systems
RDS521RL025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $290.11
Specifications

Description

Source: E. coli-derived, His37-Ser194, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.4 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Entrez Gene ID: 29197
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18.4 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
29197
IL-18/IL-1F4
25μg
Rat