missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-10 Recombinant Protein

$1,041.55 - $1,041.55

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25325
Name IL-10
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Molecular Weight Quantity Price Quantity  
522RL025 R&D Systems
RDS522RL025
Lyophilized from a 0.2μm filtered solution in Tris-HCl and NaCl with BSA as a carrier protein. 19 kDa (monomer) 25μg Each for $1,041.55
522RL025CF R&D Systems
RDS522RL025CF
Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl. 19 kDa 25μg Each for $1,041.55
Specifications

Description

Source: E. coli-derived, Ser19-Asn178
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 19 kDa
Formulation: Supplied as a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 25325
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
IL-10
Yes
25325
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat