Please login to your online account to display your discounted pricing

R&D Systems Rat IL-10 (Cys167Tyr) Recombinant Protein

$389.76 - $389.76

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25325
Molecular Weight 18.7 kDa (monomer)
Name IL-10 (Cys167Tyr)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

547RL010

 
r&d systems
RDS547RL010
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl with BSA as a carrier protein. Each for $389.76

547RL010CF

 
r&d systems
RDS547RL010CF
Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl. Each for $389.76
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25325
Molecular Weight 18.7 kDa (monomer)
Name IL-10 (Cys167Tyr)
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 10μg
Recombinant Yes
Source Rat

Source: E. coli-derived, Ser19-Asn178 (Cys167Tyr)
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18.7 kDa (monomer)
Formulation: Lyophilized from a 0.2μm filtered solution in NaH2PO4 and NaCl.
Entrez Gene ID: 25325
Regulatory Status: RUO