missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-1 beta/IL-1F2 Recombinant Protein

$376.73 - $1,101.51

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24494
Molecular Weight 17 kDa
Name IL-1 beta/IL-1F2
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
501RL010 R&D Systems
RDS501RL010
Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000 with BSA as a carrier protein. 10μg Each for $376.73
501RL010CF R&D Systems
RDS501RL010CF
Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000. 10μg Each for $376.73
501RL050 R&D Systems
RDS501RL050
Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000 with BSA as a carrier protein. 50μg Each for $1,101.51
501RL050CF R&D Systems
RDS501RL050CF
Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000. 50μg Each for $1,101.51
Specifications

Description

Source: E. coli-derived, Val117-Ser268, with an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 17 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MES, NaCl, EDTA, DTT and PEG 8000.
Entrez Gene ID: 24494
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
17 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
24494
IL-1 beta/IL-1F2
Yes