missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat IL-1 alpha/IL-1F1 Recombinant Protein

$337.62 - $520.12

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 24493
Molecular Weight 18 kDa
Name IL-1 alpha/IL-1F1
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
Catalog Number Mfr. No. Formulation Quantity Price Quantity  
500RL005 R&D Systems
RDS500RL005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $337.62
500RL025 R&D Systems
RDS500RL025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $520.12
500RL005CF R&D Systems
RDS500RL005CF
Lyophilized from a 0.2μm filtered solution in PBS. 5μg Each for $337.62
500RL025CF R&D Systems
RDS500RL025CF
Lyophilized from a 0.2μm filtered solution in PBS. 25μg Each for $520.12
Specifications

Description

Source: E. coli-derived, Ser115-Ser270
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 18 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 24493
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
18 kDa
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Rat
24493
IL-1 alpha/IL-1F1
Yes