Please login to your online account to display your discounted pricing

R&D Systems Rat GM-CSF Recombinant Protein

$164.61 - $407.05

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 116630
Molecular Weight 14.7 kDa
Name GM-CSF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs
Products
Catalog Number Mfr. No. Formulation Quantity Price Quantity    

518GM005

 
r&d systems
RDS518GM005
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 5μg Each for $164.61

518GM005CF

 
r&d systems
RDS518GM005CF
Supplied as a 0.2μm filtered solution in PBS. 5μg Each for $164.61

518GM025

 
r&d systems
RDS518GM025
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 25μg Each for $407.05

518GM025CF

 
r&d systems
RDS518GM025CF
Supplied as a 0.2μm filtered solution in PBS. 25μg Each for $407.05
Description & Specifications

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 116630
Molecular Weight 14.7 kDa
Name GM-CSF
Purity 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Recombinant Yes
Source Rat

Source: E. coli-derived, Ala1-Lys127, with and without an N-terminal Met
Purity: 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 14.7 kDa
Formulation: Supplied as a 0.2μm filtered solution in PBS.
Entrez Gene ID: 116630
Regulatory Status: RUO