Please login to your online account to display your discounted pricing

R&D Systems Rat FGF basic Recombinant Protein

$330.49 - $330.49

Specifications

Conjugate Unlabeled
Molecular Weight 16.2 kDa
Name FGF basic
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
View More Specs
Products
Catalog Number Mfr. No. Formulation Price Quantity    

3339FB025CF

 
r&d systems
RDS3339FB025CF
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT. Each for $330.49

3339FB025

 
r&d systems
RDS3339FB025
Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT with BSA as a carrier protein. Each for $330.49
Description & Specifications

Specifications

Conjugate Unlabeled
Molecular Weight 16.2 kDa
Name FGF basic
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Quantity 25μg
Recombinant Yes
Source Rat

Source: E. coli-derived, Ala11-Ser154
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 16.2 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in MOPS, Na2SO4, EDTA and DTT.
Regulatory Status: RUO