missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

R&D Systems Rat Fas Ligand/TNFSF6 Recombinant Protein

$441.90 - $441.90

Specifications

Conjugate Unlabeled
Gene ID (Entrez) 25385
Molecular Weight 21 kDa
Name Fas Ligand/TNFSF6
Purity 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Formulation Price Quantity  
Catalog Number Mfr. No. Formulation Price Quantity  
1614FL025CF R&D Systems
RDS1614FL025CF
Lyophilized from a 0.2μm filtered solution in PBS. Each for $441.90
Specifications

Description

Source: Mouse myeloma cell line, NS0-derived, Leu104-Leu278, with an N-terminal 7-His tag
Purity: 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Molecular Weight: 21 kDa
Formulation: Lyophilized from a 0.2μm filtered solution in PBS.
Entrez Gene ID: 25385
Regulatory Status: RUO

Description & Specifications

Specifications

Unlabeled
21 kDa
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Yes
25385
Fas Ligand/TNFSF6
25μg
Rat